missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl L-(-)-Lactate 98.0+%, TCI America™
Supplier: TCI America L0163100G
Specifications
| Methyl L-(-)-Lactate | |
| Colorless | |
| C4H8O3 | |
| 100 g | |
| methyl s---lactate, methyl l-lactate, s-methyl lactate, --methyl l-lactate, s-lactic acid methyl ester, methyl 2s-2-hydroxypropanoate, --methyl lactate, methyl s-2-hydroxypropionate, propanoic acid, 2-hydroxy-, methyl ester, 2s, --lactic acid methyl ester | |
| COC(=O)[C@H](C)O | |
| 104.11 | |
| CHEBI:83222 | |
| ≥98.0% (GC) |
| 27871-49-4 | |
| 140°C | |
| MFCD00064265 | |
| 3272 | |
| LPEKGGXMPWTOCB-VKHMYHEASA-N | |
| methyl (2S)-2-hydroxypropanoate | |
| 94386 | |
| 104.11 | |
| Liquid |
Chemical Identifiers
| 27871-49-4 | |
| 104.11 | |
| LPEKGGXMPWTOCB-VKHMYHEASA-N | |
| 94386 | |
| methyl (2S)-2-hydroxypropanoate |
| C4H8O3 | |
| MFCD00064265 | |
| methyl s---lactate, methyl l-lactate, s-methyl lactate, --methyl l-lactate, s-lactic acid methyl ester, methyl 2s-2-hydroxypropanoate, --methyl lactate, methyl s-2-hydroxypropionate, propanoic acid, 2-hydroxy-, methyl ester, 2s, --lactic acid methyl ester | |
| CHEBI:83222 | |
| COC(=O)[C@H](C)O |
Safety and Handling
EINECSNumber : (2)-1370
TSCA : Yes