missing translation for 'onlineSavingsMsg'
Learn More

Methyl isoxazole-5-carboxylate, 97%

Catalog No. p-7028051
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2516903 1 g
AAB2516906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2516903 Supplier Thermo Scientific Chemicals Supplier No. B2516903

CAS: 15055-81-9 | C5H5NO3 | 127.099 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15055-81-9
Molecular Formula C5H5NO3
Molecular Weight (g/mol) 127.099
MDL Number MFCD01570794
InChI Key ILPCPKZAAQXHKL-UHFFFAOYSA-N
Synonym methyl isoxazole-5-carboxylate, 5-isoxazolecarboxylic acid, methyl ester, isoxazole-5-carboxylic acid methyl ester, acmc-209wih, methyl 5-isoxazolcarboxylate, methyl 5-isoxazolcarboxylate;, methylisoxazole-5-carboxylate, 5-methoxycarbonyl-1,2-oxazol, 5-isoxazol-carbonsaeure-methylester;, isoxazol-5-carbonsaeure-methylester;
PubChem CID 2736892
IUPAC Name methyl 1,2-oxazole-5-carboxylate
SMILES COC(=O)C1=CC=NO1

Specifications

Melting Point 48°C to 50°C
Quantity 1 g
Formula Weight 127.1
Percent Purity 97%
Chemical Name or Material Methyl isoxazole-5-carboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only