missing translation for 'onlineSavingsMsg'
Learn More

Methyl Isovanillate 98.0+%, TCI America™

Catalog No. p-7133089
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22055G 5 g
M220525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22055G Supplier TCI America Supplier No. M22055G

Chemical Identifiers

CAS 6702-50-7
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD01321262
InChI Key QXOXUEFXRSIYSW-UHFFFAOYSA-N
Synonym methyl isovanillate, methyl 3-hydroxy-4-methoxybenzenecarboxylate, 3-hydroxy-4-methoxybenzoic acid methyl ester, isovanillic acid methyl ester, methyl-3-hydroxy-4-methoxybenzoate, methyl 3-hydroxy-4-methoxy-benzoate, benzoic acid, 3-hydroxy-4-methoxy-, methyl ester, methyl 3-hydroxy-4-methoxy benzoate, isovamllic acid methyl ester, ksc496e5l
PubChem CID 4056967
IUPAC Name methyl 3-hydroxy-4-methoxybenzoate
SMILES COC(=O)C1=CC(O)=C(OC)C=C1

Specifications

Melting Point 64°C
Quantity 5 g
Formula Weight 182.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Isovanillate