missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Isovalerate 98.0+%, TCI America™
$45.57 - $186.07
Chemical Identifiers
| CAS | 556-24-1 |
|---|---|
| Molecular Formula | C6H12O2 |
| Molecular Weight (g/mol) | 116.16 |
| MDL Number | MFCD00042866 |
| InChI Key | OQAGVSWESNCJJT-UHFFFAOYSA-N |
| Synonym | methyl isovalerate, methyl isopentanoate, methyl 3-methylbutyrate, isovaleric acid, methyl ester, butanoic acid, 3-methyl-, methyl ester, methyl isovalerianate, methylisovalerate, isovaleric acid methyl ester, methyl isovalerate natural, fema no. 2753 |
| PubChem CID | 11160 |
| IUPAC Name | methyl 3-methylbutanoate |
| SMILES | CC(C)CC(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
I019825ML
|
TCI America
I019825ML |
25 mL |
Each for $45.57
|
|
|||||
|
I0198500ML
|
TCI America
I0198500ML |
500 mL |
Each for $186.07
|
|
|||||
Chemical Identifiers
| 556-24-1 | |
| 116.16 | |
| OQAGVSWESNCJJT-UHFFFAOYSA-N | |
| 11160 | |
| CC(C)CC(=O)OC |
| C6H12O2 | |
| MFCD00042866 | |
| methyl isovalerate, methyl isopentanoate, methyl 3-methylbutyrate, isovaleric acid, methyl ester, butanoic acid, 3-methyl-, methyl ester, methyl isovalerianate, methylisovalerate, isovaleric acid methyl ester, methyl isovalerate natural, fema no. 2753 | |
| methyl 3-methylbutanoate |
Specifications
| 556-24-1 | |
| 118°C | |
| MFCD00042866 | |
| 2400 | |
| OQAGVSWESNCJJT-UHFFFAOYSA-N | |
| methyl 3-methylbutanoate | |
| 11160 | |
| ≥98.0% (GC) | |
| Methyl Isovalerate |
| Colorless | |
| C6H12O2 | |
| 25 mL | |
| methyl isovalerate, methyl isopentanoate, methyl 3-methylbutyrate, isovaleric acid, methyl ester, butanoic acid, 3-methyl-, methyl ester, methyl isovalerianate, methylisovalerate, isovaleric acid methyl ester, methyl isovalerate natural, fema no. 2753 | |
| CC(C)CC(=O)OC | |
| 116.16 | |
| 116.16 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0776
RTECSNumber : NY1510000
TSCA : Yes