missing translation for 'onlineSavingsMsg'
Learn More

Methyl Isovalerate 98.0+%, TCI America™

Catalog No. I0198500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0198500ML 500 mL
I019825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0198500ML Supplier TCI America Supplier No. I0198500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 556-24-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00042866
InChI Key OQAGVSWESNCJJT-UHFFFAOYSA-N
Synonym methyl isovalerate, methyl isopentanoate, methyl 3-methylbutyrate, isovaleric acid, methyl ester, butanoic acid, 3-methyl-, methyl ester, methyl isovalerianate, methylisovalerate, isovaleric acid methyl ester, methyl isovalerate natural, fema no. 2753
PubChem CID 11160
IUPAC Name methyl 3-methylbutanoate
SMILES CC(C)CC(=O)OC

Specifications

Color Colorless
Boiling Point 118°C
Quantity 500 mL
UN Number 2400
Formula Weight 116.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Isovalerate
EINECSNumber (2)-0776
RTECSNumber NY1510000
TSCA Yes