Learn More
Methyl isopropyl carbonate, 97%
CAS: 51729-83-0 | C5H10O3 | 118.13 g/mol
Supplier: Thermo Scientific Chemicals L1531406
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Methyl isopropyl carbonate | |
| 0.975 | |
| 27°C (80°F) | |
| 1.383 | |
| 5 g | |
| 1744735 | |
| RCIJMMSZBQEWKW-UHFFFAOYSA-N | |
| methyl propan-2-yl carbonate | |
| 7021530 | |
| 97% |
| 51729-83-0 | |
| 116°C to 118°C | |
| C5H10O3 | |
| MFCD01075689 | |
| UN3272 | |
| methyl isopropyl carbonate, isopropyl methyl carbonate, acmc-20akf7, ch3 c o och ch3 2, carbonic acid methylisopropyl ester | |
| COC(=O)OC(C)C | |
| 118.13 | |
| 118.13 |
Chemical Identifiers
| 51729-83-0 | |
| 118.13 | |
| RCIJMMSZBQEWKW-UHFFFAOYSA-N | |
| 7021530 | |
| COC(=O)OC(C)C |
| C5H10O3 | |
| MFCD01075689 | |
| methyl isopropyl carbonate, isopropyl methyl carbonate, acmc-20akf7, ch3 c o och ch3 2, carbonic acid methylisopropyl ester | |
| methyl propan-2-yl carbonate |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only