missing translation for 'onlineSavingsMsg'
Learn More

Methyl isopropyl carbonate, 97%

Catalog No. AAL1531403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1531403 1 g
AAL1531406 5 g
AAL1531414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1531403 Supplier Thermo Scientific Chemicals Supplier No. L1531403

CAS: 51729-83-0 | C5H10O3 | 118.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51729-83-0
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD01075689
InChI Key RCIJMMSZBQEWKW-UHFFFAOYSA-N
Synonym methyl isopropyl carbonate, isopropyl methyl carbonate, acmc-20akf7, ch3 c o och ch3 2, carbonic acid methylisopropyl ester
PubChem CID 7021530
IUPAC Name methyl propan-2-yl carbonate
SMILES COC(=O)OC(C)C

Specifications

Density 0.975
Boiling Point 116°C to 118°C
Flash Point 27°C (80°F)
Refractive Index 1.383
Quantity 1 g
UN Number UN3272
Beilstein 1744735
Formula Weight 118.13
Percent Purity 97%
Chemical Name or Material Methyl isopropyl carbonate
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only