missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Isocyanoacetate 97.0+%, TCI America™
$125.13 - $423.54
Chemical Identifiers
| CAS | 39687-95-1 |
|---|---|
| Molecular Formula | C4H7NO2 |
| Molecular Weight (g/mol) | 101.11 |
| MDL Number | MFCD00000006 |
| InChI Key | ZRGVTTCCASYOFN-UHFFFAOYSA-N |
| Synonym | methyl isocyanoacetate, isocyanoacetic acid methyl ester, methylisocyanoacetate, isocyano-acetic acid methyl ester, acetic acid,2-isocyano-, methyl ester, methyl-isocyanoacetate, methyl isocyano-acetate, acmc-20akf8, methyl alpha-isocyanoacetate, ksc581o3d |
| PubChem CID | 547815 |
| IUPAC Name | methyl 2-(methylideneamino)acetate |
| SMILES | COC(=O)CN=C |
Chemical Identifiers
| 39687-95-1 | |
| 101.11 | |
| ZRGVTTCCASYOFN-UHFFFAOYSA-N | |
| 547815 | |
| COC(=O)CN=C |
| C4H7NO2 | |
| MFCD00000006 | |
| methyl isocyanoacetate, isocyanoacetic acid methyl ester, methylisocyanoacetate, isocyano-acetic acid methyl ester, acetic acid,2-isocyano-, methyl ester, methyl-isocyanoacetate, methyl isocyano-acetate, acmc-20akf8, methyl alpha-isocyanoacetate, ksc581o3d | |
| methyl 2-(methylideneamino)acetate |
Specifications
| 39687-95-1 | |
| C4H7NO2 | |
| 1 g | |
| methyl isocyanoacetate, isocyanoacetic acid methyl ester, methylisocyanoacetate, isocyano-acetic acid methyl ester, acetic acid,2-isocyano-, methyl ester, methyl-isocyanoacetate, methyl isocyano-acetate, acmc-20akf8, methyl alpha-isocyanoacetate, ksc581o3d | |
| COC(=O)CN=C | |
| 101.11 | |
| 99.09 | |
| Liquid |
| 75°C | |
| MFCD00000006 | |
| 2922 | |
| ZRGVTTCCASYOFN-UHFFFAOYSA-N | |
| methyl 2-(methylideneamino)acetate | |
| 547815 | |
| ≥97.0% (GC) | |
| Methyl Isocyanoacetate |
Safety and Handling
TSCA : No
Recommended Storage : Freezer