accessibility menu, dialog, popup

UserName

H3COONCH2

CAS RN 39687-95-1

IUPAC Name: methyl 2-(methylideneamino)acetate
Synonyms: methyl 2-(methylideneamino)acetate
Molecular Weight (g/mol): 101.11
Molecular Formula: C4H7NO2
InChi Key: ZRGVTTCCASYOFN-UHFFFAOYSA-N
SMILES: COC(=O)CN=C
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (4)
  • (2)