missing translation for 'onlineSavingsMsg'
Learn More

Methyl Isocyanoacetate 97.0+%, TCI America™

Catalog No. M25491G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25491G 1 g
M25495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25491G Supplier TCI America Supplier No. M25491G
Only null left

Chemical Identifiers

CAS 39687-95-1
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
MDL Number MFCD00000006
InChI Key ZRGVTTCCASYOFN-UHFFFAOYSA-N
Synonym methyl isocyanoacetate, isocyanoacetic acid methyl ester, methylisocyanoacetate, isocyano-acetic acid methyl ester, acetic acid,2-isocyano-, methyl ester, methyl-isocyanoacetate, methyl isocyano-acetate, acmc-20akf8, methyl alpha-isocyanoacetate, ksc581o3d
PubChem CID 547815
IUPAC Name methyl 2-(methylideneamino)acetate
SMILES COC(=O)CN=C

Specifications

Boiling Point 75°C
Quantity 1 g
UN Number 2922
Formula Weight 99.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Isocyanoacetate
TSCA No
Recommended Storage Freezer