missing translation for 'onlineSavingsMsg'
Learn More

Methyl isocyanoacetate, 95%, Tech.

Catalog No. p-3736925
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC219000050 5 g Glass bottle
AC219000250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC219000050 Supplier Thermo Scientific Chemicals Supplier No. 219000050
Only null left

CAS: 39687-95-1 | C4H7NO2 | 101.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39687-95-1
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
MDL Number MFCD00000006
InChI Key ZRGVTTCCASYOFN-UHFFFAOYSA-N
Synonym methyl isocyanoacetate, isocyanoacetic acid methyl ester, methylisocyanoacetate, isocyano-acetic acid methyl ester, acetic acid,2-isocyano-, methyl ester, methyl-isocyanoacetate, methyl isocyano-acetate, acmc-20akf8, methyl alpha-isocyanoacetate, ksc581o3d
PubChem CID 547815
IUPAC Name methyl 2-isocyanoacetate
SMILES COC(=O)CN=C

Specifications

Color Yellow to Brown
Density 1.0900g/mL
Boiling Point 75°C to 76°C (10.0 mmHg)
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Refractive Index 1.4161 to 1.4191
Linear Formula CNCH2CO2CH3
Quantity 5 g
Specific Gravity 1.09
Formula Weight 99.09
Percent Purity 95%
Grade Technical
Physical Form Liquid
Chemical Name or Material Methyl isocyanoacetate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breath
EINECSNumber 254-593-8

RUO – Research Use Only