missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl isocyanoacetate, 95%, Tech.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Yellow to Brown |
| Density | 1.0900g/mL |
| Boiling Point | 75°C to 76°C (10.0 mmHg) |
| Flash Point | 84°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 95% |
| Packaging | Glass bottle |
| Refractive Index | 1.4161 to 1.4191 |
| Linear Formula | CNCH2CO2CH3 |
| Quantity | 25 g |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful in contact with skin. Harmful if inhaled. Causes severe skin burns and eye damage. Harmful if swallowed. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breath |
| EINECSNumber | 254-593-8 |
RUO – Research Use Only