missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Iso-Butyl Ketone, Purified (4-Methyl-2-pentanone; MIBK), Reagents
Methyl Iso-Butyl Ketone, Purified (4-Methyl-2-pentanone; MIBK), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C2250600500C
Specifications
| Methyl Iso-Butyl Ketone, Purified (4-Methyl-2-pentanone, MIBK) | |
| 108-10-1 | |
| 114°C to 117°C | |
| Glass Bottle | |
| 0.798 | |
| 4-Methyl-2-pentanone, MIBK, Isopropylacetone, Hexone | |
| CC(C)CC(C)=O | |
| 100.16 |
| -84°C | |
| Colorless | |
| 500 mL | |
| Liquid | |
| C6H12O | |
| NTIZESTWPVYFNL-UHFFFAOYSA-N | |
| 4-methylpentan-2-one | |
| Reagent |
Chemical Identifiers
| 108-10-1 | |
| 100.16 | |
| 4-Methyl-2-pentanone, MIBK, Isopropylacetone, Hexone | |
| CC(C)CC(C)=O |
| C6H12O | |
| NTIZESTWPVYFNL-UHFFFAOYSA-N | |
| 4-methylpentan-2-one |