missing translation for 'onlineSavingsMsg'
Learn More

Methyl Iso-Butyl Ketone, Purified (4-Methyl-2-pentanone; MIBK), Reagents

Catalog No. p-200021122
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337173 20 L
01337172 4 L
01337171 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337173 Supplier Labchem Inc Supplier No. C225060020E6
Only null left

Methyl Iso-Butyl Ketone, Purified (4-Methyl-2-pentanone; MIBK), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
Synonym 4-Methyl-2-pentanone, MIBK, Isopropylacetone, Hexone
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(C)=O

Specifications

Melting Point -84°C
Color Colorless
Boiling Point 114°C to 117°C
Quantity 20 L
Packaging Steel Pail
Physical Form Liquid
Specific Gravity 0.798
Grade Reagent
Chemical Name or Material Methyl Iso-Butyl Ketone, Purified (4-Methyl-2-pentanone, MIBK)