missing translation for 'onlineSavingsMsg'
Learn More

Methyl indole-7-carboxylate, 97%

Catalog No. AAH6672906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6672906 5 g
AAH6672903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6672906 Supplier Thermo Scientific Chemicals Supplier No. H6672906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 93247-78-0 | C10H9NO2 | 175.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 93247-78-0
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00211064
InChI Key FTLOEULOTNVCGF-UHFFFAOYSA-N
Synonym methyl indole-7-carboxylate, 1h-indole-7-carboxylic acid methyl ester, indole-7-carboxylic acid methyl ester, methylindole-7-carboxylate, indole-7-carboxylicacidmethylester, methyl-indole-7-carboxylate, methyl 7-indolecarboxylate, 7-indolecarboxylic acid methyl ester, 1h-indole-7-carboxylic acid,methyl ester, 1h-indole-7-carboxylic acid, methyl ester
PubChem CID 676694
IUPAC Name methyl 1H-indole-7-carboxylate
SMILES COC(=O)C1=C2NC=CC2=CC=C1

Specifications

Quantity 5 g
Beilstein 4402170
Formula Weight 175.19
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl indole-7-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only