accessibility menu, dialog, popup

UserName

CAS RN 93247-78-0

IUPAC Name: methyl 1H-indole-7-carboxylate
Synonyms: methyl 1H-indole-7-carboxylate
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: FTLOEULOTNVCGF-UHFFFAOYSA-N
SMILES: COC(=O)C1=C2NC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results