missing translation for 'onlineSavingsMsg'
Learn More

Methyl indole-6-carboxylate, 97%, Thermo Scientific™

Chemical Identifiers

CAS 50820-65-0
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00211063
InChI Key AYYOZKHMSABVRP-UHFFFAOYSA-N
Synonym methyl indole-6-carboxylate, indole-6-carboxylic acid methyl ester, methylindole-6-carboxylate, 1h-indole-6-carboxylic acid, methyl ester, 1h-indole-6-carboxylic acid methyl ester, indole-7-carboxylate, 6-carbomethoxyindole, indole-6-carboxylate, pubchem1696, 6-methoxycarbonylindole
PubChem CID 639844
IUPAC Name methyl 1H-indole-6-carboxylate
SMILES COC(=O)C1=CC2=C(C=C1)C=CN2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC397930010
View Documents Promotion Details
N/A 1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC397930050
View Documents Promotion Details
N/A 5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

50820-65-0
175.19
AYYOZKHMSABVRP-UHFFFAOYSA-N
639844
COC(=O)C1=CC2=C(C=C1)C=CN2
C10H9NO2
MFCD00211063
methyl indole-6-carboxylate, indole-6-carboxylic acid methyl ester, methylindole-6-carboxylate, 1h-indole-6-carboxylic acid, methyl ester, 1h-indole-6-carboxylic acid methyl ester, indole-7-carboxylate, 6-carbomethoxyindole, indole-6-carboxylate, pubchem1696, 6-methoxycarbonylindole
methyl 1H-indole-6-carboxylate

Specifications

50820-65-0
96% min. (GC)
MFCD00211063
AYYOZKHMSABVRP-UHFFFAOYSA-N
methyl 1H-indole-6-carboxylate
639844
97%
Authentic
C10H9NO2
methyl indole-6-carboxylate, indole-6-carboxylic acid methyl ester, methylindole-6-carboxylate, 1h-indole-6-carboxylic acid, methyl ester, 1h-indole-6-carboxylic acid methyl ester, indole-7-carboxylate, 6-carbomethoxyindole, indole-6-carboxylate, pubchem1696, 6-methoxycarbonylindole
COC(=O)C1=CC2=C(C=C1)C=CN2
175.19
175.19
Methyl indole-6-carboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only