missing translation for 'onlineSavingsMsg'
Learn More

Methyl indole-6-carboxylate, 97%

Catalog No. AC397930010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397930010 1 g Glass bottle
AC397930050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397930010 Supplier Thermo Scientific Chemicals Supplier No. 397930010

CAS: 50820-65-0 | C10H9NO2 | 175.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50820-65-0
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00211063
InChI Key AYYOZKHMSABVRP-UHFFFAOYSA-N
Synonym methyl indole-6-carboxylate, indole-6-carboxylic acid methyl ester, methylindole-6-carboxylate, 1h-indole-6-carboxylic acid, methyl ester, 1h-indole-6-carboxylic acid methyl ester, indole-7-carboxylate, 6-carbomethoxyindole, indole-6-carboxylate, pubchem1696, 6-methoxycarbonylindole
PubChem CID 639844
IUPAC Name methyl 1H-indole-6-carboxylate
SMILES COC(=O)C1=CC2=C(C=C1)C=CN2

Specifications

Melting Point 76°C to 83°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 175.19
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material Methyl indole-6-carboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw

RUO – Research Use Only