missing translation for 'onlineSavingsMsg'
Learn More

Methyl Indole-3-acetate 98.0+%, TCI America™

Catalog No. M26055G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26055G 5 g
M260525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26055G Supplier TCI America Supplier No. M26055G

Chemical Identifiers

CAS 1912-33-0
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.214
MDL Number MFCD00022749
InChI Key KTHADMDGDNYQRX-UHFFFAOYSA-N
Synonym Indole-3-acetic Acid Methyl Ester, IAA Methyl Ester
PubChem CID 74706
ChEBI CHEBI:72782
IUPAC Name methyl 2-(1H-indol-3-yl)acetate
SMILES COC(=O)CC1=CNC2=CC=CC=C21

Specifications

Melting Point 51°C
Boiling Point 160°C
Quantity 5 g
Formula Weight 189.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl Indole-3-acetate
TSCA No
Recommended Storage Refrigerator