accessibility menu, dialog, popup

UserName

H3COOHN

CAS RN 1912-33-0

IUPAC Name: methyl 2-(1H-indol-3-yl)acetate
Synonyms: methyl 2-(1H-indol-3-yl)acetate
Molecular Weight (g/mol): 189.21
Molecular Formula: C11H11NO2
InChi Key: KTHADMDGDNYQRX-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CNC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results