missing translation for 'onlineSavingsMsg'
Learn More

Methyl Indazole-6-carboxylate 97.0+%, TCI America™

Catalog No. M28231G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28231G 1 g
M28235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28231G Supplier TCI America Supplier No. M28231G

Chemical Identifiers

CAS 170487-40-8
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.175
MDL Number MFCD07371612
InChI Key TUSICEWIXLMXEY-UHFFFAOYSA-N
Synonym 1h-indazole-6-carboxylic acid methyl ester, methyl indazole-6-carboxylate, indazole-6-carboxylic acid methyl ester, 6-1h indazole carboxylic acid methyl ester, 1h-indazole-6-carboxylic acid, methyl ester, pubchem18064, methyl indazole-6carboxylate, ksc494o4p, methyl-1h-indazole-6-carboxylate, 2h-indazole-6-carboxylic acid methyl ester
PubChem CID 286535
IUPAC Name methyl 1H-indazole-6-carboxylate
SMILES COC(=O)C1=CC2=C(C=C1)C=NN2

Specifications

Melting Point 144°C
Quantity 1 g
Formula Weight 176.18
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Indazole-6-carboxylate