accessibility menu, dialog, popup

UserName

CH3OONHN

CAS RN 170487-40-8

IUPAC Name: methyl 1H-indazole-6-carboxylate
Synonyms: methyl 1H-indazole-6-carboxylate
Molecular Weight (g/mol): 176.18
Molecular Formula: C9H8N2O2
InChi Key: TUSICEWIXLMXEY-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C2C=NNC2=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)