missing translation for 'onlineSavingsMsg'
Learn More

Methyl imidazo[1,2-a]pyridine-7-carboxylate, 95%, Thermo Scientific Chemicals

Catalog No. p-7032893
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3531003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3531003 Supplier Thermo Scientific Chemicals Supplier No. H3531003

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86718-01-6
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.18
MDL Number MFCD09910350
InChI Key KYHRVKMXYXBEQN-UHFFFAOYSA-N
Synonym methyl imidazo 1,2-a pyridine-7-carboxylate, imidazo 1,2-a pyridine-7-carboxylic acid methyl ester, 7-methoxycarbonyl imidazo 1,2-a pyridine, methylimidazo 1,2-a pyridine-7-carboxylate, methyl h-imidazo 1,2-a pyridine-7-carboxylate, imidazo 1,2-a pyridine-7-carboxylic acid, methyl ester, pubchem21685, methylimidazoapyridinecarboxylate, methyl imidazo-1,2-a pyridine-7-carboxylate, 7-imidazo 1,2-a pyridinecarboxylic acid methyl ester
PubChem CID 15096964
IUPAC Name methyl imidazo[1,2-a]pyridine-7-carboxylate
SMILES COC(=O)C1=CC2=NC=CN2C=C1

Specifications

Melting Point 133°C to 135°C
Quantity 1 g
Formula Weight 176.18
Percent Purity 95%
Chemical Name or Material Methyl imidazo[1,2-a]pyridine-7-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only