accessibility menu, dialog, popup

UserName

CAS RN 86718-01-6

IUPAC Name: methyl imidazo[1,2-a]pyridine-7-carboxylate
Synonyms: methyl imidazo[1,2-a]pyridine-7-carboxylate
Molecular Weight (g/mol): 176.18
Molecular Formula: C9H8N2O2
InChi Key: KYHRVKMXYXBEQN-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC2=NC=CN2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results