missing translation for 'onlineSavingsMsg'
Learn More

Methyl hydrogen succinate, 95%

CAS: 3878-55-5 | C5H8O4 | 132.115 g/mol

$157.56 - $424.79

Chemical Identifiers

CAS 3878-55-5
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.115
MDL Number MFCD00002788
InChI Key JDRMYOQETPMYQX-UHFFFAOYSA-N
Synonym monomethyl succinate, succinic acid monomethyl ester, methyl hydrogen succinate, mono-methyl succinate, butanedioic acid, monomethyl ester, 3-carbomethoxypropanoic acid, succinic acid, monomethyl ester, unii-ya2v724s0a, mono-methyl hydrogen succinate, butanedioic acid monomethyl ester
PubChem CID 77487
ChEBI CHEBI:75146
IUPAC Name 4-methoxy-4-oxobutanoic acid
SMILES COC(=O)CCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAH5972214
View Documents Promotion Details
N/A 25 g
Each for $157.56
Only null left
 
AAH5972222
View Documents Promotion Details
N/A 100 g
Each for $424.79
Only null left
 
Description

Description

It could react with 1,2,3-trimethoxy-benzene, and obtain the 4-oxo-4-(2,3,4-trimethoxy-phenyl)-butyric acid methyl ester. This reaction needs the reagent of PPA. Butanedioic Acid 1-Methyl Ester is an intermediate in the synthesis of agonist to both GHB and GABAB neural receptors, exhibiting neurotransmitter-like effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It could react with 1,2,3-trimethoxy-benzene, and obtain the 4-oxo-4-(2,3,4-trimethoxy-phenyl)-butyric acid methyl ester. This reaction needs the reagent of PPA. Butanedioic Acid 1-Methyl Ester is an intermediate in the synthesis of agonist to both GHB and GABAB neural receptors, exhibiting neurotransmitter-like effects.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

3878-55-5
132.115
JDRMYOQETPMYQX-UHFFFAOYSA-N
77487
4-methoxy-4-oxobutanoic acid
C5H8O4
MFCD00002788
monomethyl succinate, succinic acid monomethyl ester, methyl hydrogen succinate, mono-methyl succinate, butanedioic acid, monomethyl ester, 3-carbomethoxypropanoic acid, succinic acid, monomethyl ester, unii-ya2v724s0a, mono-methyl hydrogen succinate, butanedioic acid monomethyl ester
CHEBI:75146
COC(=O)CCC(=O)O

Specifications

3878-55-5
151°C (20 mmHg)
CH3OCOCH2CH2CO2H
1722669
Slightly soluble in water.
COC(=O)CCC(=O)O
132.115
CHEBI:75146
95%
54°C to 57°C
C5H8O4
MFCD00002788
monomethyl succinate, succinic acid monomethyl ester, methyl hydrogen succinate, mono-methyl succinate, butanedioic acid, monomethyl ester, 3-carbomethoxypropanoic acid, succinic acid, monomethyl ester, unii-ya2v724s0a, mono-methyl hydrogen succinate, butanedioic acid monomethyl ester
JDRMYOQETPMYQX-UHFFFAOYSA-N
4-methoxy-4-oxobutanoic acid
77487
132.11
Methyl hydrogen succinate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 223-408-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only