missing translation for 'onlineSavingsMsg'
Learn More

Methyl hydrogen phthalate, 97%

Catalog No. AAA1478009
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1478009 10 g
AAA1478018 50 g
AAA1478030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1478009 Supplier Thermo Scientific Chemicals Supplier No. A1478009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4376-18-5 | C9H8O4 | 180.16 g/mol

Methyl hydrogen phthalate is used as a phthalate ester metabolite, linked to precocious puberty.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl hydrogen phthalate is used as a phthalate ester metabolite, linked to precocious puberty.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 4376-18-5
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
MDL Number MFCD00002466
InChI Key FNJSWIPFHMKRAT-UHFFFAOYSA-N
Synonym 2-methoxycarbonyl benzoic acid, methyl hydrogen phthalate, monomethyl phthalate, mono-methyl phthalate, phthalic acid monomethyl ester, o-methoxycarbonyl benzoic acid, monomethyl 1,2-benzenedicarboxylate, phthalic acid, monomethyl ester, monomethylphthalate, unii-2stt6d18jr
PubChem CID 20392
IUPAC Name 2-methoxycarbonylbenzoic acid
SMILES COC(=O)C1=CC=CC=C1C(O)=O

Specifications

Melting Point 81°C to 84°C
Quantity 10 g
Beilstein 2048766
Solubility Information Insoluble in water.
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material Methyl hydrogen phthalate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-476-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only