missing translation for 'onlineSavingsMsg'
Learn More

Methyl Heptanoate 99.5+%, TCI America™

Catalog No. s03055ml
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03055ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03055ML Supplier TCI America Supplier No. S03055ML
Only null left

Chemical Identifiers

CAS 106-73-0
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00009537
InChI Key XNCNNDVCAUWAIT-UHFFFAOYSA-N
Synonym methyl enanthate, heptanoic acid, methyl ester, methyl oenanthylate, methyl heptoate, methyl heptylate, heptanoic acid methyl ester, methyl n-heptylate, enanthic acid methyl ester, methyl heptylate natural, methyl n-heptanoate
PubChem CID 7826
IUPAC Name methyl heptanoate
SMILES CCCCCCC(=O)OC

Specifications

Color Colorless
Boiling Point 172°C
Quantity 5 mL
UN Number 1993
Formula Weight 144.21
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Heptanoate [Standard Material for GC]
EINECSNumber (2)-0782
RTECSNumber MJ2297500
TSCA Yes