missing translation for 'onlineSavingsMsg'
Learn More

Methyl heptanoate, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020221
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020221 1 mL
111018696 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020221 Supplier Merck Emd Millipore Supplier No. 752181ML
Only null left

Methyl heptanoate is a fatty acid ester prepared by treating heptanoic acid with methyl alcohol in the presence of hydrochloric acid or sulfuric acid. It is commonly used as a flavoring agent in food products.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Packaging Glass Bottle
CAS 106-73-0
Density 0.880 g/mL (at 20°C)
Flash Point 52°C - Closed Cup
Shipping Condition Ambient
Physical Form Neat
Linear Formula C8H16O2
MDL Number MFCD00009537
Boiling Point 171°C to 172°C
Synonym Heptanoic acid methyl ester; Methyl enanthate
Type Analytical Standard
Quantity 1 mL
Formula Weight 144.21
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less