missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Heptafluorobutyrate 97.0+%, TCI America™
$164.03 - $456.70
Chemical Identifiers
| CAS | 356-24-1 |
|---|---|
| Molecular Formula | C5H3F7O2 |
| Molecular Weight (g/mol) | 228.066 |
| MDL Number | MFCD00000433 |
| InChI Key | MRPUVAKBXDBGJQ-UHFFFAOYSA-N |
| Synonym | methyl heptafluorobutyrate, methyl heptafluorobutanoate, methyl perfluorobutyrate, methylheptafluorobutyrate, butanoic acid, heptafluoro-, methyl ester, heptafluorobutyric acid methyl ester, butyric acid, heptafluoro-, methyl ester, butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, methyl ester, heptafluorobutyryl methyl ester, methyl perfluorbutanoate |
| PubChem CID | 67741 |
| IUPAC Name | methyl 2,2,3,3,4,4,4-heptafluorobutanoate |
| SMILES | COC(=O)C(C(C(F)(F)F)(F)F)(F)F |
Chemical Identifiers
| 356-24-1 | |
| 228.066 | |
| MRPUVAKBXDBGJQ-UHFFFAOYSA-N | |
| 67741 | |
| COC(=O)C(C(C(F)(F)F)(F)F)(F)F |
| C5H3F7O2 | |
| MFCD00000433 | |
| methyl heptafluorobutyrate, methyl heptafluorobutanoate, methyl perfluorobutyrate, methylheptafluorobutyrate, butanoic acid, heptafluoro-, methyl ester, heptafluorobutyric acid methyl ester, butyric acid, heptafluoro-, methyl ester, butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, methyl ester, heptafluorobutyryl methyl ester, methyl perfluorbutanoate | |
| methyl 2,2,3,3,4,4,4-heptafluorobutanoate |
Specifications
| 356-24-1 | |
| 81°C | |
| MFCD00000433 | |
| 3272 | |
| MRPUVAKBXDBGJQ-UHFFFAOYSA-N | |
| methyl 2,2,3,3,4,4,4-heptafluorobutanoate | |
| 67741 | |
| ≥97.0% (GC) | |
| Methyl Heptafluorobutyrate |
| Colorless | |
| C5H3F7O2 | |
| 5 g | |
| methyl heptafluorobutyrate, methyl heptafluorobutanoate, methyl perfluorobutyrate, methylheptafluorobutyrate, butanoic acid, heptafluoro-, methyl ester, heptafluorobutyric acid methyl ester, butyric acid, heptafluoro-, methyl ester, butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, methyl ester, heptafluorobutyryl methyl ester, methyl perfluorbutanoate | |
| COC(=O)C(C(C(F)(F)F)(F)F)(F)F | |
| 228.066 | |
| 228.07 | |
| Liquid |
Safety and Handling
TSCA : Yes