missing translation for 'onlineSavingsMsg'
Learn More

Methyl Heptadecanoate 98.0+%, TCI America™

Catalog No. p-7130612
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05665G 5 g
H056625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05665G Supplier TCI America Supplier No. H05665G

Chemical Identifiers

CAS 1731-92-6
Molecular Formula C18H36O2
Molecular Weight (g/mol) 284.484
MDL Number MFCD00009001
InChI Key HUEBIMLTDXKIPR-UHFFFAOYSA-N
Synonym methyl margarate, heptadecanoic acid, methyl ester, margaric acid methyl ester, heptadecanoic acid methyl ester, unii-41pzk7h0kl, 41pzk7h0kl, heptadecanoic acid,methyl ester, n-heptadecanoic acid methyl ester, methylheptadecanoate, formyl heptadecanoate
PubChem CID 15609
IUPAC Name methyl heptadecanoate
SMILES CCCCCCCCCCCCCCCCC(=O)OC

Specifications

Color White
Quantity 5 g
Formula Weight 284.48
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl Heptadecanoate