missing translation for 'onlineSavingsMsg'
Learn More

Methyl heneicosanoate, ≥98.5% (GC), MilliporeSigma™ Supelco™

Numéro de catalogue. 111014865
Modifier l'affichage
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111014865 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111014865 Fournisseur Merck Emd Millipore Code fournisseur 515351G
Il en reste null

Spécifications

CAS 6064-90-0
Flash Point >113°C
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)19 COOCH3
MDL Number MFCD00009345
Melting Point 48°C to 50°C; 48°C to 50°C
Boiling Point 207°C (4 mmHg)
Synonym Heneicosanoic acid methyl ester
Type Analytical Standard
Quantity 1 g
Formula Weight 340.58
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Afficher plus de résultats Afficher moins de résultats