Learn More
Thermo Scientific Chemicals Methyl Green, zinc chloride
CAS: 7114-03-6 | C27H35Cl4N3Zn | 608.78 g/mol
$220.66 - $711.42
Chemical Identifiers
| CAS | 7114-03-6 |
|---|---|
| Molecular Formula | C27H35Cl4N3Zn |
| Molecular Weight (g/mol) | 608.78 |
| MDL Number | MFCD00151094 |
| InChI Key | ZKOGXCSOLMZRAZ-UHFFFAOYSA-J |
| Synonym | C.I. 42590 |
| PubChem CID | 131675247 |
| IUPAC Name | zinc;[4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-ethyl-dimethylazanium;bromide;trichloride |
| SMILES | CC[N+](C)(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-].[Cl-].[Cl-].[Zn+2].[Br-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6150914
|
Thermo Scientific Chemicals
J6150914 |
25 g |
Each for $220.66
|
|
|||||
|
AAJ6150922
|
Thermo Scientific Chemicals
J6150922 |
100 g |
Each for $711.42
|
|
|||||
Description
Methyl Green is a DNA stain used as a tracking dye in acid buffers for electrophoresis. It helps differentiate bacteria and is also a tissue stain for nucleoli and mitochondria in tissues. It is used with Bismarck Brown frequently.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl Green is a DNA stain used as a tracking dye in acid buffers for electrophoresis. It helps differentiate bacteria and is also a tissue stain for nucleoli and mitochondria in tissues. It is used with Bismarck Brown frequently.
Solubility
Soluble in water, ethanol or methanol
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 7114-03-6 | |
| 608.78 | |
| ZKOGXCSOLMZRAZ-UHFFFAOYSA-J | |
| 131675247 | |
| CC[N+](C)(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-].[Cl-].[Cl-].[Zn+2].[Br-] |
| C27H35Cl4N3Zn | |
| MFCD00151094 | |
| C.I. 42590 | |
| zinc;[4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-ethyl-dimethylazanium;bromide;trichloride |
Specifications
| >300°C | |
| C27H35Cl4N3Zn | |
| 3914106 | |
| Soluble in water,ethanol or methanol | |
| CC[N+](C)(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-].[Cl-].[Cl-].[Zn+2].[Br-] | |
| 608.78 | |
| 653.24 | |
| 25 g | |
| Methyl Green, zinc chloride |
| 7114-03-6 | |
| MFCD00151094 | |
| C.I. 42590 | |
| ZKOGXCSOLMZRAZ-UHFFFAOYSA-J | |
| zinc;[4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-ethyl-dimethylazanium;bromide;trichloride | |
| 131675247 | |
| Green | |
| Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 230-415-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only