Learn More
Thermo Scientific Chemicals Methyl Green, zinc chloride
CAS: 7114-03-6 | C27H35Cl4N3Zn | 608.78 g/mol
Supplier: Thermo Scientific Chemicals J6150922
Description
Methyl Green is a DNA stain used as a tracking dye in acid buffers for electrophoresis. It helps differentiate bacteria and is also a tissue stain for nucleoli and mitochondria in tissues. It is used with Bismarck Brown frequently.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl Green is a DNA stain used as a tracking dye in acid buffers for electrophoresis. It helps differentiate bacteria and is also a tissue stain for nucleoli and mitochondria in tissues. It is used with Bismarck Brown frequently.
Solubility
Soluble in water, ethanol or methanol
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| Methyl Green, zinc chloride | |
| 7114-03-6 | |
| MFCD00151094 | |
| C.I. 42590 | |
| ZKOGXCSOLMZRAZ-UHFFFAOYSA-J | |
| zinc;[4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-ethyl-dimethylazanium;bromide;trichloride | |
| 131675247 | |
| Green | |
| Powder |
| >300°C | |
| C27H35Cl4N3Zn | |
| 3914106 | |
| Soluble in water,ethanol or methanol | |
| CC[N+](C)(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-].[Cl-].[Cl-].[Zn+2].[Br-] | |
| 608.78 | |
| 653.24 | |
| 100 g |
Chemical Identifiers
| 7114-03-6 | |
| 608.78 | |
| ZKOGXCSOLMZRAZ-UHFFFAOYSA-J | |
| 131675247 | |
| CC[N+](C)(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-].[Cl-].[Cl-].[Zn+2].[Br-] |
| C27H35Cl4N3Zn | |
| MFCD00151094 | |
| C.I. 42590 | |
| zinc;[4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-ethyl-dimethylazanium;bromide;trichloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 230-415-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only