missing translation for 'onlineSavingsMsg'
Learn More

Methyl Fumaraldehydate 96.0+%, TCI America™

Catalog No. F0351100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0351100MG 100 mg
F03511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0351100MG Supplier TCI America Supplier No. F0351100MG

Chemical Identifiers

CAS 5837-72-9
Molecular Formula C5H6O3
Molecular Weight (g/mol) 114.1
MDL Number MFCD00191651
InChI Key CRBJVPSOOMDSPT-NSCUHMNNSA-N
Synonym 3-(Methoxycarbonyl)-2-propenal, Fumaraldehydic Acid Methyl Ester, 3-(Methoxycarbonyl)acrylaldehyde
PubChem CID 5364747
IUPAC Name methyl (E)-4-oxobut-2-enoate
SMILES COC(=O)C=CC=O

Specifications

Melting Point 40°C
Color Yellow
Boiling Point 88°C
Quantity 100 mg
Formula Weight 114.10
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Fumaraldehydate
TSCA No
Recommended Storage Refrigerator