accessibility menu, dialog, popup

UserName

CH3OOO(E/Z)

CAS RN 5837-72-9

IUPAC Name: methyl 4-oxobut-2-enoate
Synonyms: methyl 4-oxobut-2-enoate
Molecular Weight (g/mol): 114.1
Molecular Formula: C5H6O3
InChi Key: CRBJVPSOOMDSPT-UHFFFAOYSA-N
SMILES: COC(=O)C=CC=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results