missing translation for 'onlineSavingsMsg'
Learn More

Methyl Ethyl Ketone/Toluene, 90g/60g, Reagents

Catalog No. 01333419
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01333419 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01333419 Supplier Labchem Inc Supplier No. CT1442004C
Only null left

Methyl Ethyl Ketone/Toluene, 90g/60g, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 78-93-3 , 108-88-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
Synonym Methyl Ethyl Ketone/Toluol
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

Color Colorless
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.8293
Grade Reagent
Chemical Name or Material Methyl Ethyl Ketone/Toluene, 90 g/60 g