missing translation for 'onlineSavingsMsg'
Learn More

Methyl eicosanoate, 98+%

Catalog No. AA4239909
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4239909 10 g
AA4239904 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4239909 Supplier Thermo Scientific Chemicals Supplier No. 04239909

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1120-28-1 | C21H42O2 | 326.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1120-28-1
Molecular Formula C21H42O2
Molecular Weight (g/mol) 326.57
MDL Number MFCD00009014
InChI Key QGBRLVONZXHAKJ-UHFFFAOYSA-N
Synonym methyl arachidate, eicosanoic acid, methyl ester, arachidic acid methyl ester, methyl eicosanoate, methyl arachisate, arachidic acid methylester, eicosanoic acid methyl ester, methylarachidate, methyl aracidate
PubChem CID 14259
IUPAC Name methyl icosanoate
SMILES CCCCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 48°C
Color White
Boiling Point 215°C (10 mmHg)
Flash Point >110°C (230°F)
Quantity 10 g
Solubility Information Soluble in hot alcohol. Sparingly Soluble in water.
Formula Weight 326.56
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl eicosanoate
EINECSNumber 214-304-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only