missing translation for 'onlineSavingsMsg'
Learn More

Methyl (E)-4-Chloro-3-methoxy-2-butenoate 95.0+%, TCI America™

Catalog No. p-7124924
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13405G 5 g
C134025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13405G Supplier TCI America Supplier No. C13405G

Chemical Identifiers

CAS 110104-60-4
Molecular Formula C6H9ClO3
Molecular Weight (g/mol) 164.585
MDL Number MFCD00071562
InChI Key JNYMRXDQVPIONI-HWKANZROSA-N
Synonym e-methyl 4-chloro-3-methoxybut-2-enoate, methyl e-4-chloro-3-methoxy-2-butenoate, e-4-chloro-3-methoxy-2-butenoic acid methyl ester, methyl 4-chloro-3-methoxy-2-e-butenoate, 4-chloro-3-methoxy-2-e-butenoic acid methyl ester, 4-chloro-3-methoxy-2 e-butenoic acid methyl ester, methyl 2e-4-chloro-3-methoxybut-2-enoate, hedl`lxpbdee jzzafh, methyl e-4-chloro-3-methoxy-2-butanoate
PubChem CID 6364655
IUPAC Name methyl (E)-4-chloro-3-methoxybut-2-enoate
SMILES COC(=CC(=O)OC)CCl

Specifications

Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 164.59
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl (E)-4-Chloro-3-methoxy-2-butenoate