accessibility menu, dialog, popup

UserName

CH3OOClOH3C(E)

CAS RN 110104-60-4

IUPAC Name: methyl (2E)-4-chloro-3-methoxybut-2-enoate
Synonyms: methyl (2E)-4-chloro-3-methoxybut-2-enoate
Molecular Weight (g/mol): 164.59
Molecular Formula: C6H9ClO3
InChi Key: JNYMRXDQVPIONI-HWKANZROSA-N
SMILES: COC(=O)\C=C(/CCl)OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results