missing translation for 'onlineSavingsMsg'
Learn More

Methyl dodecanoate, 99%

CAS: 111-82-0 | C13H26O2 | 214.349 g/mol

Supplier:  Thermo Scientific Chemicals A1249214

 View more versions of this product

Catalog No. AAA1249214


Only null left
Explore available promotions

Description

Description

Methyl dodecanoate used in production of biodiesel. It has been used in selective synthesis of the secondary amide surfactant, N-methyl lauroylethanolamide. It is also utilized as emollients, perfuming agents skin conditioning.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl dodecanoate used in production of biodiesel. It has been used in selective synthesis of the secondary amide surfactant, N-methyl lauroylethanolamide. It is also utilized as emollients, perfuming agents skin conditioning.

Solubility
Not miscible in water. Soluble in alcohol, dipropylene glycol.

Notes
Store at room temperature. Ensure good ventilation.
Specifications

Specifications

Methyl dodecanoate
4°C to 5°C
261°C to 262°C
99%
1.4325
25 g
methyl laurate, lauric acid methyl ester, dodecanoic acid, methyl ester, methyl laurinate, methyl dodecylate, methyl n-dodecanoate, uniphat a40, metholene 2296, lauric acid, methyl ester, stepan c40
UQDUPQYQJKYHQI-UHFFFAOYSA-N
methyl dodecanoate
8139
214.35
Liquid
111-82-0
0.87
125°C (257°F)
C13H26O2
MFCD00008966
1767780
Not miscible in water. Soluble in alcohol,dipropylene glycol.
CCCCCCCCCCCC(=O)OC
214.349
CHEBI:87494
99%

Chemical Identifiers

111-82-0
214.349
UQDUPQYQJKYHQI-UHFFFAOYSA-N
8139
methyl dodecanoate
C13H26O2
MFCD00008966
methyl laurate, lauric acid methyl ester, dodecanoic acid, methyl ester, methyl laurinate, methyl dodecylate, methyl n-dodecanoate, uniphat a40, metholene 2296, lauric acid, methyl ester, stepan c40
CHEBI:87494
CCCCCCCCCCCC(=O)OC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 203-911-3

RTECSNumber : OF0670000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only