missing translation for 'onlineSavingsMsg'
Learn More

Methyl dodecanoate, 99%

Catalog No. AAA1249222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1249222 100 g
AAA1249206 5 g
AAA1249214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1249222 Supplier Thermo Scientific Chemicals Supplier No. A1249222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 111-82-0 | C13H26O2 | 214.349 g/mol

Methyl dodecanoate used in production of biodiesel. It has been used in selective synthesis of the secondary amide surfactant, N-methyl lauroylethanolamide. It is also utilized as emollients, perfuming agents skin conditioning.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl dodecanoate used in production of biodiesel. It has been used in selective synthesis of the secondary amide surfactant, N-methyl lauroylethanolamide. It is also utilized as emollients, perfuming agents skin conditioning.

Solubility
Not miscible in water. Soluble in alcohol, dipropylene glycol.

Notes
Store at room temperature. Ensure good ventilation.

Chemical Identifiers

CAS 111-82-0
Molecular Formula C13H26O2
Molecular Weight (g/mol) 214.349
MDL Number MFCD00008966
InChI Key UQDUPQYQJKYHQI-UHFFFAOYSA-N
Synonym methyl laurate, lauric acid methyl ester, dodecanoic acid, methyl ester, methyl laurinate, methyl dodecylate, methyl n-dodecanoate, uniphat a40, metholene 2296, lauric acid, methyl ester, stepan c40
PubChem CID 8139
ChEBI CHEBI:87494
IUPAC Name methyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OC

Specifications

Melting Point 4°C to 5°C
Density 0.87
Boiling Point 261°C to 262°C
Flash Point 125°C (257°F)
Assay Percent Range 99%
Refractive Index 1.4325
Quantity 100 g
Beilstein 1767780
Solubility Information Not miscible in water. Soluble in alcohol,dipropylene glycol.
Formula Weight 214.35
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl dodecanoate
Show More Show Less
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 203-911-3
RTECSNumber OF0670000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only