missing translation for 'onlineSavingsMsg'
Learn More

Methyl dodecanoate, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019390
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019390 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019390 Supplier Merck Emd Millipore Supplier No. 616895ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 111-82-0
Density 0.869 g/mL (at 20°C)
Flash Point 115.6°C
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)10 COOCH3
MDL Number MFCD00008966
Melting Point 4.5°C to 5°C
Boiling Point 261°C to 262°C
Synonym Lauric acid methyl ester; Methyl laurate
Type Analytical Standard
Quantity 5 mL
Formula Weight 214.34
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less