Learn More
Methyl disulfide, 99%
CAS: 624-92-0 | C2H6S2 | 94.19 g/mol
Supplier: Thermo Scientific Chemicals 165590025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Methyl disulfide | |
| -85.0°C | |
| 1.0625g/mL | |
| 16°C | |
| 1% max. (Methylmercaptan content) (GC) | |
| C2H6S2 | |
| (CH3)2S2 | |
| 2.5 kg | |
| 05,246; 06,217; 08,189 | |
| dimethyl disulfide, methyl disulfide, dimethyldisulfide, disulfide, dimethyl, dmds, 2,3-dithiabutane, dimethyl disulphide, methyldisulfanyl methane, methyldisulfide, methyldithiomethane | |
| WQOXQRCZOLPYPM-UHFFFAOYSA-N | |
| (methyldisulfanyl)methane | |
| 12232 | |
| CHEBI:4608 | |
| 99% |
| 624-92-0 | |
| Yellow | |
| 108.0°C to 110.0°C | |
| Authentic | |
| Glass bottle | |
| 1.5243 to 1.5263 | |
| MFCD00008561 | |
| 01, 291 | |
| 1.0625 | |
| Solubility in water: insoluble. Other solubilities: soluble in ether,ethyl alcohol,ccl4,benzene | |
| CSSC | |
| 94.19 | |
| 0.620 mPa.s (20°C) | |
| 94.19 | |
| Liquid |
Chemical Identifiers
| 624-92-0 | |
| 94.19 | |
| WQOXQRCZOLPYPM-UHFFFAOYSA-N | |
| 12232 | |
| (methyldisulfanyl)methane |
| C2H6S2 | |
| MFCD00008561 | |
| dimethyl disulfide, methyl disulfide, dimethyldisulfide, disulfide, dimethyl, dmds, 2,3-dithiabutane, dimethyl disulphide, methyldisulfanyl methane, methyldisulfide, methyldithiomethane | |
| CHEBI:4608 | |
| CSSC |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapour.
Toxic if swallowed.
Toxic if inhaled.
May cause an allergic skin reaction.
Causes serious eye irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
I
GHS Signal Word: Danger
EINECSNumber : 210-871-
RUO – Research Use Only