missing translation for 'onlineSavingsMsg'
Learn More

Methyl disulfide, 99%, AcroSeal™

Catalog No. AC430821000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430821000 100 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC430821000 Supplier Thermo Scientific Chemicals Supplier No. 430821000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 624-92-0 | C2H6S2 | 94.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 624-92-0
Molecular Formula C2H6S2
Molecular Weight (g/mol) 94.19
MDL Number MFCD00008561
InChI Key WQOXQRCZOLPYPM-UHFFFAOYSA-N
Synonym dimethyl disulfide, methyl disulfide, dimethyldisulfide, disulfide, dimethyl, dmds, 2,3-dithiabutane, dimethyl disulphide, methyldisulfanyl methane, methyldisulfide, methyldithiomethane
PubChem CID 12232
ChEBI CHEBI:4608
IUPAC Name (methyldisulfanyl)methane
SMILES CSSC

Specifications

Melting Point -85.0°C
Color Yellow
Density 1.0625g/mL
Boiling Point 108.0°C to 110.0°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 1% max. Methylmercaptan content (GC)
Packaging AcroSeal™ Glass bottle
Refractive Index 1.5243 to 1.5263
Linear Formula (CH3)2S2
Quantity 100 mL
Specific Gravity 1.0625
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether,ethyl alcohol,ccl4,benzene
Viscosity 0.620 mPa.s (20°C)
Formula Weight 94.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl disulfide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Toxic if swallowed.
Toxic if inhaled.
May cause an allergic skin reaction.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
I
EINECSNumber 210-871-

RUO – Research Use Only