missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl diphenylphosphinite, 98+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1.078 |
| Boiling Point | 148°C to 150°C (6 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.604 |
| Quantity | 1 g |
| Beilstein | 1640480 |
| Sensitivity | Air and moisture sensitive |
| Formula Weight | 216.22 |
| Percent Purity | ≥98% |
| Chemical Name or Material | Methyl diphenylphosphinite |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 223-683-9 |
| TSCA | No |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only