accessibility menu, dialog, popup

UserName

CH3OP

CAS RN 4020-99-9

IUPAC Name: methyl diphenylphosphinite
Synonyms: methyl diphenylphosphinite
Molecular Weight (g/mol): 216.22
Molecular Formula: C13H13OP
InChi Key: OAADXJFIBNEPLY-UHFFFAOYSA-N
SMILES: COP(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)