missing translation for 'onlineSavingsMsg'
Learn More

Methyl diphenylphosphinite, 98+%

Catalog No. AAA1155503
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1155503 1 g
AAA1155514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1155503 Supplier Thermo Scientific Chemicals Supplier No. A1155503

CAS: 4020-99-9 | C13H13OP | 216.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4020-99-9
Molecular Formula C13H13OP
Molecular Weight (g/mol) 216.22
MDL Number MFCD00048025
InChI Key OAADXJFIBNEPLY-UHFFFAOYSA-N
Synonym methoxydiphenylphosphine, methyl diphenylphosphinite, diphenylphosphinous acid methyl ester, phosphinous acid, diphenyl-, methyl ester, methoxy diphenyl phosphane, pubchem6470, acmc-209jbw, diphenyl methoxy phosphine
PubChem CID 77636
IUPAC Name methoxy(diphenyl)phosphane
SMILES COP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Density 1.078
Boiling Point 148°C to 150°C (6 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.604
Quantity 1 g
Beilstein 1640480
Sensitivity Air and moisture sensitive
Formula Weight 216.22
Percent Purity ≥98%
Chemical Name or Material Methyl diphenylphosphinite
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 223-683-9
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only