missing translation for 'onlineSavingsMsg'
Learn More

Methyl Diphenylacetate 98.0+%, TCI America™

Catalog No. D288825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D288825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D288825G Supplier TCI America Supplier No. D288825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3469-00-9
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.28
MDL Number MFCD00025869
InChI Key AORIUCNKPVHMTN-UHFFFAOYSA-N
Synonym methyl diphenylacetate, diphenylacetic acid, methyl ester, acetic acid, diphenyl-, methyl ester, benzeneacetic acid, .alpha.-phenyl-, methyl ester, methyldiphenylacetate, cbmicro_020304, acmc-209i9m, c6h5 2chcooch3, diphenylacetic acid methyl ester
PubChem CID 77019
IUPAC Name methyl 2,2-diphenylacetate
SMILES COC(=O)C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 61°C
Quantity 25 g
Formula Weight 226.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Diphenylacetate