missing translation for 'onlineSavingsMsg'
Learn More

Methyl diphenylacetate, 98%

Catalog No. p-7027717
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
5 g
Catalog No. Quantity
AAB2469406 5 g
AAB2469414 25 g
AAB2469422 100 g
3 options

Catalog No. AAB2469406

Supplier: Thermo Scientific Chemicals B2469406

Only null left

CAS: 3469-00-9 | C15H14O2 | 226.28 g/mol

Methyl diphenylacetate is used as a chemical, organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl diphenylacetate is used as a chemical, organic intermediate.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3469-00-9
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.28
MDL Number MFCD00025869
InChI Key AORIUCNKPVHMTN-UHFFFAOYSA-N
Synonym methyl diphenylacetate, diphenylacetic acid, methyl ester, acetic acid, diphenyl-, methyl ester, benzeneacetic acid, .alpha.-phenyl-, methyl ester, methyldiphenylacetate, cbmicro_020304, acmc-209i9m, c6h5 2chcooch3, diphenylacetic acid methyl ester
PubChem CID 77019
IUPAC Name methyl 2,2-diphenylacetate
SMILES COC(=O)C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 59°C to 61°C
Quantity 5 g
Solubility Information Soluble in methanol.
Formula Weight 226.28
Percent Purity 98%
Chemical Name or Material Methyl diphenylacetate
EINECSNumber 222-431-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only