missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl diphenylacetate, 98%
Description
Methyl diphenylacetate is used as a chemical, organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl diphenylacetate is used as a chemical, organic intermediate.
Solubility
Soluble in methanol.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Methyl diphenylacetate is used as a chemical, organic intermediate.
Solubility
Soluble in methanol.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 59°C to 61°C |
| Quantity | 25 g |
| Solubility Information | Soluble in methanol. |
| Formula Weight | 226.28 |
| Percent Purity | 98% |
| Chemical Name or Material | Methyl diphenylacetate |
Safety and Handling
| EINECSNumber | 222-431-5 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only